Learn More
Invitrogen™ SSBP1 Polyclonal Antibody

Description
Immunogen sequence: LQVLRQFVRH ESETTTSLVL ERSLNRVHLL GRVGQDPVLR QVEGKNPVTI FSLATNEMWR SGDSEVYQLG DVSQKTTWHR ISVFRPGLRD VAYQYVKKGS RIYLEGKIDY GEYMDKNNVR RQATTIIADN IIFLSDQTKE Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 89%.
Specifications
Specifications
| Antigen | SSBP1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | Ssbp1 |
| Gene Accession No. | P28042, Q04837, Q9CYR0 |
| Gene Alias | 2810480P10Rik; G630031O20Rik; mtDBP; mtSSB; Mt-SSB; PWP1-interacting protein 17; single strand DNA-binding protein P16; single stranded DNA binding protein 1; single-stranded DNA binding protein 1; single-stranded DNA binding protein 1, mitochondrial; single-stranded DNA-binding protein, mitochondrial; SOSS-B1; Ssbp; Ssbp1 |
| Gene Symbols | Ssbp1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.