Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals SSC buffer (20X)

Catalog No. AAJ60839K7
Change view
Click to view available options
Quantity:
1 L
2 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60839K7 4 L
AAJ60839K2 1 L
AAJ60839K3 2 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ60839K7 Supplier Thermo Scientific Chemicals Supplier No. J60839K7
Add to Cart

missing translation for 'validMainframeMsg'

Suitable for preparing nucleic acids for hybridization, hybridization buffer for northern and southern blots, washing buffer for northern and southern blots. Saline-Sodium Citrate Buffer (SSC) is used as a hybridization buffer in immunoblotting and DNA Microarray protocols.

SSC buffer is suitable for preparing nucleic acids for hybridization, hybridization buffer for northern and southern blots, washing buffer for northern and southern blots. Saline-Sodium Citrate Buffer (SSC) is used as a hybridization buffer in immunoblotting and DNA Microarray protocols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Physical Form Liquid
Quantity 4 L

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.