Learn More
Thermo Scientific Chemicals SSC Buffer (20X) PROMO
Catalog No. J60839K7PM
Change view
Click to view available options
4 L
Catalog No.
J60839K7PM 4 L
1 options
Catalog No. J60839K7PM Supplier Thermo Scientific Chemicals Supplier No. J60839K7PR
Suitable for preparing nucleic acids for hybridization, hybridization buffer for northern and southern blots, washing buffer for northern and southern blots. Saline-Sodium Citrate Buffer (SSC) is used as a hybridization buffer in immunoblotting and DNA Microarray protocols.
TRUSTED_SUSTAINABILITY
Specifications
Physical Form Liquid
Quantity 4 L