Learn More
Invitrogen™ SSEA1 Monoclonal Antibody (MEM-158), FITC

Description
This antibody reacts with CD15, a cell membrane molecule 3-fucosyl-N-acetyllactosamine (3-FAL) strongly expressed on the surface of granulocytes, monocytes, macrophages, mast cells; it is also present on Langerhans cells and some myeloid precursors cells. This antibody will not cross-react with porcine, bovine or sheep.
Specifications
Specifications
| Antigen | SSEA1 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | MEM-158 |
| Conjugate | FITC |
| Formulation | TBS with 0.2% BSA and 15mM sodium azide; pH 8.0 |
| Gene | FUT4 |
| Gene Accession No. | P22083 |
| Gene Alias | FUTIV; Lewis X; SSEA-1 |
| Gene Symbols | FUT4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.