Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SSEA3 Monoclonal Antibody (eBioMC-631 (MC-631)), Biotin, eBioscience™
GREENER_CHOICE

Catalog No. p-7178477
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-122-22 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-122-22 Supplier Invitrogen™ Supplier No. 13883382
Only null left
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Description: The eBioMC-631 (MC-631) monoclonal antibody reacts with the carbohydrate moiety galactosylgloboside on Stage-specific embryonic antigen-3 (SSEA-3). Expression is restricted to the surface of human teratocarcinoma stem cells (EC), human embryonic germ cells (EG) and human embryonic stem cells (ES) and down regulated following differentiation. Applications Reported: This eBioMC-631 (MC-631) antibody has been reported for use in flow cytometric analysis. Applications Tested: This eBioMC-631 (MC-631) antibody has been tested by flow cytometric analysis of the 2102Ep cell line. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Filtration: 0.2 μm post-manufacturing filtered.

SSEA-3 (stage-specific embryonic antigen 3) is a cell surface antigen, expressed along with SSEA-4, TRA-1-60 and TRA-1-81 in embryonic stem cells, embryonal carcinoma cells, and induced pluripotent stem cells (iPS). In human cells, these surface markers are down-regulated during the differentiation process, while SSEA1 is absent in undifferentiated human stem cells and upregulated on the cell surface after retinoic acid mediated differentiation. Conversely, in mouse stem cells, SSEA-1 expression decreases with differentiation while SSEA-3 and SSEA-4 expression increase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SSEA3
Applications Flow Cytometry
Classification Monoclonal
Clone eBioMC-631 (MC-631)
Concentration 0.5 mg/mL
Conjugate Biotin
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Alias SSEA-3; stage-specific embryonic antigen 3
Host Species Rat
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.