Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SSEA3 Monoclonal Antibody (MC-631)
GREENER_CHOICE

Catalog No. PIMA1020 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
20 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA1020 100 μg
PIMA1020X 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIMA1020 Supplier Invitrogen™ Supplier No. MA1020
  • $559.50
    $409.65 / Each of 1

    Save $149.85

    27% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rat Monoclonal Antibody

SSEA3 Monoclonal Antibody for Flow, ICC/IF

SSEA-3 (stage-specific embryonic antigen 3) is a cell surface antigen, expressed along with SSEA-4, TRA-1-60 and TRA-1-81 in embryonic stem cells, embryonal carcinoma cells, and induced pluripotent stem cells (iPS). In human cells, these surface markers are down-regulated during the differentiation process, while SSEA1 is absent in undifferentiated human stem cells and upregulated on the cell surface after retinoic acid mediated differentiation. Conversely, in mouse stem cells, SSEA-1 expression decreases with differentiation while SSEA-3 and SSEA-4 expression increase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SSEA3
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone MC-631
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation Potassium phosphate with 0.15M NaCl, 1mg/mL BSA and 0.05% sodium azide
Gene Alias SSEA-3; stage-specific embryonic antigen 3
Host Species Rat
Immunogen Fischer rat immunized with 4-8 cell stage mouse embryos.
Purification Method Affinity chromatography - MBP
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.