Learn More
Description
MA1-020-PE has been successfully used in ICC/IF and flow cytometry applications on human samples.
Specifications
Specifications
| Antigen | SSEA3 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | MC-631 |
| Concentration | 1 mg/mL |
| Conjugate | PE |
| Formulation | PBS with 1mg/mL BSA and 0.05% sodium azide |
| Gene Alias | SSEA-3; stage-specific embryonic antigen 3 |
| Host Species | Rat |
| Immunogen | Fischer rat immunized with 4-8 cell stage mouse embryos. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
