Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SSEA4 Monoclonal Antibody (MC-813-70)
GREENER_CHOICE

Mouse Monoclonal Antibody

$235.87 - $543.84

Specifications

Antigen SSEA4
Clone MC-813-70
Concentration 1 mg/mL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Applications Flow Cytometry, Immunocytochemistry
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PIMA1021
GREENER_CHOICE
View Documents
Invitrogen™
MA1021
100 μg
Each for $543.84
Only null left
Add to Cart
 
PIMA1021X
View Documents
Invitrogen™
MA1021X
20 μL
Each for $235.87
Only null left
Add to Cart
 
Description

Description

SSEA4 Monoclonal Antibody for Flow, ICC/IF

SSEA-4 (Stage-specific embryonic antigen 4) is a cell surface antigen, expressed along with SSEA-3, TRA-1-60 and TRA-1-81 in embryonic stem cells, embryonal carcinoma cells and induced pluripotent stem cells (iPS). In human cells, these surface markers are down-regulated during the differentiation process, while SSEA-1 is absent in undifferentiated human stem cells and upregulated on the cell surface after retinoic acid mediated differentiation. Conversely, in mouse stem cells, SSEA-1 expression decreases with differentiation while SSEA-3 and SSEA-4 expression increase.
Specifications

Specifications

SSEA4
1 mg/mL
Flow Cytometry, Immunocytochemistry
Unconjugated
Mouse
RUO
Potassium phosphate with 0.15M NaCl, 1mg/mL BSA and 0.05% sodium azide
human embryonal carcinoma cell line 2102Ep.
Antibody
MC-813-70
-20°C, Avoid Freeze/Thaw Cycles
Monoclonal
Liquid
IgG3
Human
SSEA-4; stage-specific embryonic antigen 4
Primary
Protein A
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.