Learn More
Description
Immunofluorescence and flow cytometric analysis of MA1-144-D488 shows positive staining of SSEA-5 positive pluripotent cells and cultured human embryonal carcinoma NCCIT cells.
Specifications
Specifications
| Antigen | SSEA5 |
| Applications | Flow Cytometry, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 8E11 |
| Concentration | 1 mg/mL |
| Conjugate | DyLight 488 |
| Formulation | PBS with proprietary stabilizer and 0.02% sodium azide |
| Gene Alias | SSEA-5 |
| Host Species | Mouse |
| Immunogen | Undifferentiated H9 human embryonic stem cells. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
