Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SuperScript™ III First-Strand Synthesis SuperMix

Catalog No. 18080400
Encompass_Preferred
Click to view available options
Quantity:
50 rxns

Catalog No. 18-080-400

Supplier: Invitrogen™ 18080400

Only null left
Add to Cart
Add to Cart

Includes

2X Reaction Mix (500μL); SuperScript III Enzyme Mix (100μL); Oligo(dT)20 (50μL total provided at 50μM); Random hexamers (50μL total provided at 50ng/μL); Annealing buffer (50μL)

The SuperScript III First-Strand Synthesis SuperMix is an optimized formulation for first-strand cDNA synthesis from purified poly(A)+ or total RNA. RNA targets from 100 bp to >12 kb can be detected with this system.

The SuperScript III First-Strand Synthesis SuperMix is an optimized formulation for first-strand cDNA synthesis from purified poly(A)+ or total RNA. RNA targets from 100 bp to >12 kb can be detected with this system. The amount of starting material can vary from 0.1 pg to 5 μg of total RNA.

This kit includes SuperScript III/RNaseOUT enzyme mix, 2X First-Strand reaction mix, and an annealing buffer. SuperScript III Reverse Transcriptase is a version of MMLV RT that has been engineered to reduce RNase H activity and provide increased thermal stability. The enzyme can be used to synthesize cDNA at a temperature range of 45–55°C, providing increased specificity, higher yields of cDNA, and more full-length product than many previous reverse transcriptases.

Using SuperScript III First-Strand Synthesis SuperMix
SuperScript III RT is not significantly inhibited by ribosomal and transfer RNA and can be used to synthesize cDNA from total RNA. RNaseOUT Recombinant RNase Inhibitor is included in the enzyme mix to safeguard against degradation of target RNA due to ribonuclease contamination. The 2X First-Strand reaction mix includes 10 mM MgCl2 and 1 mM of each dNTP in a buffer formulation that has been optimized for first-strand synthesis of cDNA. The annealing buffer is used in the initial template-primer annealing step. Separate tubes of oligo(dT)20 and random hexamers are also provided. cDNA synthesis can be performed using either total RNA or poly(A)+-selected RNA primed with oligo(dT), random primers, or a gene-specific primer.

Order Info

Shipping Condition: Dry ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage

• 2X Reaction Mix, 500 μL
• SuperScript™ III Enzyme Mix, 100 μL
• Oligo(dT)20, 50 μL total (50 μM)
• Random hexamers, 50 μL (50 ng/μL)
• Annealing buffer, 50 μL

Store at –20°C.

Format Kit
GC-Rich PCR Performance High
Reaction Speed 50 min.
Technique Reverse Transcription
Optimal Reaction Temperature 50°C
Primer Random Primers, Oligo dT Primers
Quantity 50 rxns
Reverse Transcriptase SuperScript III
Shipping Condition Dry Ice
For Use With (Application) Real Time PCR (qPCR)
Final Product Type First-Strand cDNA
No. of Reactions 50 Reactions
Reaction Format Master Mix
Reagent Type Reverse Transcription
Starting Material RNA
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.