Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SSTR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA3206 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA3206 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA3206 Supplier Invitrogen™ Supplier No. PA3206
  • $591.00
    $449.65 / Each of 1

    Save $141.35

    24% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

SSTR1 Polyclonal Antibody

SSTR1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen SSTR1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation whole serum with 0.02% sodium azide
Gene SSTR1
Gene Accession No. P28646, P30873
Gene Alias Gpcrrna; RNGPCRRNA; Smstr1; Smstr-1; somatostatin receptor; somatostatin receptor 1; Somatostatin receptor subtype 1; somatostatin receptor type 1; SRIF-2; SS1R; SS-1-R; SS1-R; sst1; Sst1 receptor; Sstr1
Gene Symbols SSTR1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the C-terminal amino acids E(374)SGGVFRNGTCASRISTL(391) of SSTR1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20605, 25033
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.