Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SSTR1 Recombinant Rabbit Monoclonal Antibody (18H22L9)

Catalog No. 704008
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-400-8 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 70-400-8 Supplier Invitrogen™ Supplier No. 704008
Add to Cart
Edge
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. Recombinant antibodies are produced using specific genes that code for the desired antibodies. These genes are cloned into an expression vector and expressed in vitro. The advantages of recombinant antibodies include better specificity and lot-to-lot consistency. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

Somatostatin acts to regulate numerous physiological processes by binding to and activating specific receptors in target tissues. Activation of these receptors by somatostatin, which is secreted by nerve and endocrine cells, regulates the secretion of insulin, glucagon and growth hormone, neuronal excitability in both the brain and the peripheral nervous system, and cell growth. Somatostatin receptors have been implicated in numerous diseases ranging from Alzheimer's to cancers of the gastrointestinal tract, breast, prostate, and pituitary.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SSTR1
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 18H22L9
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2-7.4
Gene SSTR1
Gene Accession No. P28646, P30872, P30873
Gene Alias Gpcrrna; RNGPCRRNA; Smstr1; Smstr-1; somatostatin receptor; somatostatin receptor 1; Somatostatin receptor subtype 1; somatostatin receptor type 1; SRIF-2; SS1R; SS-1-R; SS1-R; sst1; Sst1 receptor; Sstr1
Gene Symbols SSTR1
Host Species Rabbit
Immunogen Peptide corresponding to the aa residues 377-391 of the human SSTR1.
Purification Method Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20605, 25033, 6751
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.