Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SSTR4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA3208
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA3208 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA3208 Supplier Thermo Scientific Supplier No. PA3208
Add to Cart
Edge
Add to Cart

SSTR4 Polyclonal Antibody

SSTR4 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen SSTR4
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation whole serum with 0.02% sodium azide
Gene SSTR4
Gene Accession No. P30937, P49660
Gene Alias G-protein coupled receptor; Smstr4; somatostatin receptor; somatostatin receptor 4; Somatostatin receptor subtype 4 Rattus norvegicus Sprague-Dawley major hippocampal somatostatin receptor (SSTR4) mRNA; somatostatin receptor type 4; SS4R; SS-4-R; SS4-R; sst4; Sstr4
Gene Symbols SSTR4
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the C-terminal amino acids C(362)QQEPVQAEPGCKQVPFTKTTTF(384) of SSTR4.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20608, 25555
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.