Learn More
Description
PA3-112 detects the Somatostatin Receptor 5 in human samples. PA3-112 has been used successfully in Western blot, immunocytochemistry, and immunohistochemistry procedures. In Western blot analysis of SSTR5 transfected HEK-293 cells this antibody detects a ~54-63 kDa protein representing SSTR5. The PA3-112 immunogen is a synthetic peptide corresponding to residues Q(344) E A T P P A H R A A A N G L M Q T S K L(364) of human SSTR5.
Specifications
Specifications
| Antigen | SSTR5 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | Whole serum with 0.05% sodium azide |
| Gene | Sstr5 |
| Gene Accession No. | O08858, P35346 |
| Gene Alias | Smstr5; Somato; somatostatin receptor; somatostatin receptor 5; Somatostatin receptor subtype 5; somatostatin receptor type 5; SS5R; SS-5-R; SS5-R; sst5; Sstr5 |
| Gene Symbols | Sstr5 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
