Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SSTR5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA3209 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA3209 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA3209 Supplier Invitrogen™ Supplier No. PA3209
  • $618.00
    $461.65 / Each of 1

    Save $156.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

PA3-209 detects SSTR5 in rat and mouse samples. PA3-209 has successfully been used in Western blot, immunocytochemical, and immunohistochemical (paraffin) procedures. The PA3-209 immunogen is a synthetic peptide corresponding to the C-terminal amino acids Q(344)ATLPTRSCEANGLMQTSRI(363) of SSTR5.

Somatostatin is a multifunctional protein affecting exocrine and endocrine protein secretion, neurotransmission, and neuronal modulation and regulating cell proliferation. All these functions of Somatostatin are executed through a class of GPCR protein family members called somatostatin receptor. Till date, 6 classes of somatostatin receptors have been discovered out of which SSTR5, 364A.A protein, is a receptor subtype expressed predominantly in the pituitary and hypothalamus region of brain. Studies revealed that SSTR5 contains seven transmembrane regions and undergoes homodimerization and also heterodimerizes with SSTR1. Although the specific functions of SSTR5 are still elusive, the studies indicate that it complements SSTR2 regulation of ACTH secretion. Polymorphisms in SSTR5 gene have been observed in patients with acromegaly.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SSTR5
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation whole serum with 0.02% sodium azide
Gene Sstr5
Gene Accession No. O08858, P30938
Gene Alias Smstr5; Somato; somatostatin receptor; somatostatin receptor 5; Somatostatin receptor subtype 5; somatostatin receptor type 5; SS5R; SS-5-R; SS5-R; sst5; Sstr5
Gene Symbols Sstr5
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the C-terminal amino acids Q(344)ATLPTRSCEANGLMQTSRI(363) of SSTR5.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20609, 25354
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.