Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SSTR5 Recombinant Rabbit Monoclonal Antibody (19H17L5)

Catalog No. 704013
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-401-3 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 70-401-3 Supplier Invitrogen™ Supplier No. 704013
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

SSTR5 Recombinant Monoclonal Antibody for ICC/IF, IHC, Western Blot

Somatostatin is a multifunctional protein affecting exocrine and endocrine protein secretion, neurotransmission, and neuronal modulation and regulating cell proliferation. All these functions of Somatostatin are executed through a class of GPCR protein family members called somatostatin receptor. Till date, 6 classes of somatostatin receptors have been discovered out of which SSTR5, 364A.A protein, is a receptor subtype expressed predominantly in the pituitary and hypothalamus region of brain. Studies revealed that SSTR5 contains seven transmembrane regions and undergoes homodimerization and also heterodimerizes with SSTR1. Although the specific functions of SSTR5 are still elusive, the studies indicate that it complements SSTR2 regulation of ACTH secretion. Polymorphisms in SSTR5 gene have been observed in patients with acromegaly.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SSTR5
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 19H17L5
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2-7.4
Gene Sstr5
Gene Accession No. P35346
Gene Alias Smstr5; Somato; somatostatin receptor; somatostatin receptor 5; Somatostatin receptor subtype 5; somatostatin receptor type 5; SS5R; SS-5-R; SS5-R; sst5; Sstr5
Gene Symbols Sstr5
Host Species Rabbit
Immunogen Peptides corresponding to residues 344-364 of Human SSTR5.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6755
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.