Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SSYA10-001
GREENER_CHOICE

Catalog No. 755410
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
75-541-0 10 mg
75-545-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 75-541-0 Supplier Tocris Bioscience™ Supplier No. 7554/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent coronavirus helicase inhibitor; also inhibits human papillomavirus E6

SSYA10-001 is a potent coronavirus helicase inhibitor (IC50 values are 0.046 μM and >3 μM for unwinding and ATPase, respectively). Inhibits SARS-CoV-2 Nsp13 helicase (IC50 = 3.5 μM), blocks dsDNA and dsRNA unwinding activities of Nsp13 (IC50 values are 5.3 μM and 5.7 μM respectively). In a SARS-CoV replicon assay SSYA10-001 inhibits (+) sense but not (-) sense RNA synthesis (EC50 = 8.95 μM). Also inhibits human papillomavirus E6 (IC50 = 10 μM)

Specifications

Chemical Name or Material 2,4-Dihydro-5-[[(2-nitrophenyl)thio]methyl]-4-(2-propen-1-yl)-3 H-1,2,4-triazole-3-thione
CAS 675104-49-1
Quantity 10 mg
Target Antiviral Compounds
Molecular Formula C12H12N4O2S2
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.