Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ST 1936 oxalate, R&D Systems™
SDP

Catalog No. 433710 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-371-0 10 mg
43-375-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 43-371-0 Supplier R&D Systems Supplier No. 4337/10

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective, high affinity 5-HT6 agonist

High affinity, selective agonist of 5-HT6 receptors (Ki values are 13, 168, and 245 nM for human 5-HT6, 5-HT7 and 5-HT2B receptors respectively). Displays moderate affinity for human α2 adrenergic receptors (Ki = 300 nM). Shown to increase Ca2+ concentration and phosphorylation of Fyn kinase, and regulate the activation of ERK1/2 in cloned cells.

Chemical Identifiers

CAS 1210-81-7
Molecular Formula C15H19ClN2O4
Molecular Weight (g/mol) 326.777
InChI Key FHSPBPPTDFRVGQ-UHFFFAOYSA-N
Synonym st 1936 oxalate, 5-chloro-3-2-dimethylamino ethyl-2-methylindole oxalate
PubChem CID 90488934
IUPAC Name 2-(5-chloro-2-methyl-1H-indol-3-yl)-N,N-dimethylethanamine;oxalic acid
SMILES CC1=C(C2=C(N1)C=CC(=C2)Cl)CCN(C)C.C(=O)(C(=O)O)O

Specifications

Quantity 10 mg
Formula Weight 326.78
Percent Purity >98%
Product Line Tocris
Chemical Name or Material ST 1936 oxalate
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.