Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IKA™ ST-20 Tube
SDP

Catalog No. 01183051 Shop All IKA Works Products
Change view
Click to view available options
For Use With (Equipment):
ULTRA-TURRAX™ Tube Drive
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
01-183-051 ULTRA-TURRAX™ Tube Drive
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 01-183-051 Supplier IKA™ Supplier No. 3703000
Get a Magnetic Stirrer with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Can be used for mixing, stirring, extractions, preparation of soil sample suspensions and solubility testing for medication.

Contains: tube, stirring device and cover

Specifications

Volume Range (Metric) 2 to 15 mL
For Use With (Application) Mixing, stirring, extractions, preparation of soil sample suspensions, solubility testing for medication
For Use With (Equipment) ULTRA-TURRAX™ Tube Drive
Packaging Quantity 1 Ea.
Product Type ST-20 Tube
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.