Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ST18 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540764
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540764 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540764 Supplier Invitrogen™ Supplier No. PA540764
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: VIKKGKVSEE LMTIKLKATG GIESDEEIRH LDEEIKELNE SNLKIEADMM Sequence homology: Cow: 86%; Dog: 93%; Guinea Pig: 86%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%.

Repressor that binds to DNA sequences containing a bipartite element consisting of a direct repeat of the sequence 5'-AAAGTTT-3' separated by 2-9 nucleotides. Represses basal transcription activity from target promoters. Inhibits colony formation in cultured breast cancer cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ST18
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ST18
Gene Accession No. O60284
Gene Alias AV348974; C2-HC type zinc finger protein r-MyT3; Kiaa0535; mKIAA0535; Myt3; Neural zinc finger factor 3; neural zinc finger protein NZF-3; neural zinc finger transcription factor 3; Nzf3; NZF-3; r-MyT3; St18; ST18 C2H2C-type zinc finger transcription factor; ST18, C2H2C-type zinc finger; suppression of tumorigenicity 18; suppression of tumorigenicity 18 (breast carcinoma) (zinc finger protein); suppression of tumorigenicity 18 protein; suppression of tumorigenicity 18, zinc finger; suppression of tumorigenicity protein 18; ZC2H2C3; ZC2HC10; zinc finger protein 387; ZNF387
Gene Symbols ST18
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human ST18.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9705
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.