Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ ST2/IL-33R Antibody Pair [HRP]
SDP

Catalog No. NB397947 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
15 Plates
5 Plates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB397947 5 Plates
NB397948 15 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB397947 Supplier Novus Biologicals™ Supplier No. NBP3116585PLATES
Only null left
Add to Cart
Add to Cart

Pair [HRP]

Specifications

Assay Range 7.8 to 500 pg/mL (example only, lot dependent)
Assay Sensitivity 7.8 pg/mL (example only; lot dependent)
Conjugate HRP
Detection Method Sandwich ELISA
Immunoassay Kit Format Sandwich ELISA Kit
Product Type Sandwich ELISA
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human ST2/IL-33 R.
Target Species Human
For Use With (Application) ELISA, Sandwich ELISA
Gene ID (Entrez) 9173
Gene Symbol IL1RL1
Quantity 5 Plates
Research Discipline Angiogenesis
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym DER4ST2growth stimulation-expressed, FIT-1, IL33R, interleukin 1 receptor-like 1, interleukin 1 receptor-related protein, interleukin-1 receptor-like 1, MGC32623, Protein ST2, ST2L, ST2V, T1homolog of mouse growth stimulation-expressed
Target Human
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.