Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ST3GAL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542726
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542726 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542726 Supplier Invitrogen™ Supplier No. PA542726
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: YVFDNWLQGH GRYPSTGILS VIFSMHVCDE VDLYGFGADS KGNWHHYWEN Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.

ST3GAL1 is a type II membrane protein that catalyzes the transfer of sialic acid from CMP-sialic acid to galactose-containing substrates. It is normally found in the Golgi but can be proteolytically processed to a soluble form. Correct glycosylation of ST3GAL1 protein may be critical to its sialyltransferase activity. This protein, which is a member of glycosyltransferase family 29, can use the same acceptor substrates as does sialyltransferase 4B.The protein encoded by this gene is a type II membrane protein that catalyzes the transfer of sialic acid from CMP-sialic acid to galactose-containing substrates. The encoded protein is normally found in the Golgi but can be proteolytically processed to a soluble form. Correct glycosylation of the encoded protein may be critical to its sialyltransferase activity. This protein, which is a member of glycosyltransferase family 29, can use the same acceptor substrates as does sialyltransferase 4B. Two transcript variants encoding the same protein have been found for this gene. Other transcript variants may exist, but have not been fully characterized yet.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ST3GAL1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ST3GAL1
Gene Accession No. Q11201
Gene Alias 5330418N22Rik; AI467004; Alpha 2,3-ST 1; alpha-2,3-sialyltransferase ST3Gal I-r1; alpha-2-3-sialyltransferase; beta-galactoside alpha-2,3-sialyltransferase 1; CMP-N-acetylneuraminate: [beta-galactosidase alpha-2,3] sialytransferase; CMP-N-acetylneuraminate-beta-galactosamide-alpha-2,3-sialyltransferase 1; Gal beta1,3 GalNAc alpha2,3 sialyltransferase; Gal-beta-1,3-GalNAc-alpha-2,3-sialyltransferase; Gal-NAc6S; MGC9183; putative alpha-2,3-sialyltransferase; Sialyltransferase 4A; sialyltransferase 4A (beta-galactosidase alpha-2,3-sialytransferase); sialyltransferase 4A (beta-galactoside alpha-2,3-sialyltransferase); sialyltransferase 4A (beta-galactoside alpha-2,3-sialytransferase); sialyltransferase ST3Gal-I; Siat4; SIAT4A; SIAT4-A; siat4-r1; SIATFL; ST3 beta-galactoside alpha-2,3-sialyltransferase 1; ST3Gal I; st3Gal I.1; ST3GAL1; St3gal-1; st3gal1A; st3gal1-r1; ST3GAL2; ST3GalA; ST3GalA.1; ST3GalI; ST3GAL-I sialyltransferase; ST3GalIA; ST3GalIA,1; ST3O
Gene Symbols ST3GAL1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human ST3GAL1 (aa 254-303).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6482
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.