Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ST6GALNAC6 Mouse anti-Human, Mouse, Clone: 1A7B7, Proteintech
SDP

Catalog No. 501736642 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6642 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6642 Supplier Proteintech Group Inc Supplier No. 662091IG150UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

ST6GALNAC6 belongs to a family of sialyltransferases that modify proteins and ceramides on the cell surface to alter cell-cell or cell-extracellular matrix interactions.

Specifications

Antigen ST6GALNAC6
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 1A7B7
Concentration 0.74 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ST6GALNAC6
Gene Accession No. Q969X2, Q9JM95
Gene Alias hST6GalNAc VI, RP11 203J24.3, Sialyltransferase 7F, SIAT7 F, SIAT7F, ST6GalNAc VI, ST6GALNAC6, ST6GALNACVI
Gene Symbols St6galnac6
Host Species Mouse
Immunogen ST6GALNAC6 Fusion Protein Ag8358
Purification Method Protein G
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30815, 50935
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.