Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ST7/LRP12 Antibody [DyLight 650], Novus Biologicals™
SDP

Catalog No. AF2560W
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2560W 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AF2560W Supplier R&D Systems Supplier No. AF2560W

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Sheep Polyclonal Antibody

ST7/LRP12 Polyclonal antibody specifically detects ST7/LRP12 in Human samples. It is validated for Western Blot

Specifications

Antigen ST7/LRP12
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias DKFZp762O2113, Protein FAM4A1, Protein HELG, SEN4, subfamily A, member 1, suppression of tumorigenicity 7, suppression of tumorigenicity 7 (breast), suppressor of tumorigenicity 7 protein
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant human ST7/LRP12, Asn28-Ile488, Accession # Q9Y561
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29967
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.