Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ST8SIA1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597234
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597234 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597234 Supplier Invitrogen™ Supplier No. PA597234
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YRLPNEKEIV QGVLQQGTAW RRNQTAARAF RKQMEDCCDP AHLFAMTKMN SPMGKSMWYD GEFLYSFTID NSTYSLFPQA TPFQLPLKKC AVVGNGGILK KSGCGRQIDE ANFVMRCNLP PLSSEYTKDV GSKSQLVTAN PSIIRQRFQN LLWSRKTFVD NMKIYNHSYI YMPAFSMKTG TEPSLRVYYT LSDVGANQTV LFANPNFLRS IGKFWKSRGI HA; Positive Samples: A-549, SGC-7901; Cellular Location: Golgi apparatus membrane, Single-pass type II membrane protein.

Gangliosides are membrane-bound glycosphingolipids containing sialic acid. Ganglioside GD3 is known to be important for cell adhesion and growth of cultured malignant cells. The protein encoded by this gene is a type II membrane protein that catalyzes the transfer of sialic acid from CMP-sialic acid to GM3 to produce gangliosides GD3 and GT3. The encoded protein may be found in the Golgi apparatus and is a member of glycosyltransferase family 29. Alternatively spliced transcript variants have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ST8SIA1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 8.42 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene ST8SIA1
Gene Accession No. Q92185
Gene Alias 9330109E03Rik; alpha-2, 8-sialyltransferase; alpha-2, 8-sialytransferase; alpha-2,8-sialyltransferase; alpha-2,8-sialyltransferase 1; alpha-2,8-sialyltransferase 8A; alpha-N-acetylneuraminate: alpha-2,8-sialytransferase; alpha-N-acetylneuraminide alpha-2,8-sialyltransferase; disialoganglioside (GD3) synthase; ganglioside GD3 synthase; Ganglioside GT3 synthase; ganglioside-specific alpha-2,8-polysialyltransferase; GD3 synthase; GD3S; sialyltransferase 8; sialyltransferase 8 (alpha-2 8-sialytransferase) A; sialyltransferase 8 (alpha-N-acetylneuraminate: alpha-2,8-sialytransferase, GD3 synthase) A; Sialyltransferase 8 (alpha-N-acetylneuraminate: alpha-28-sialytransferase GD3 synthase); sialyltransferase 8 A; Sialyltransferase 8 A (alpha-N-acetylneuraminate: alpha-28-sialytransferase GD3 synthase) GenBank no: U53883; sialyltransferase 8A; sialyltransferase 8A (alpha-N-acetylneuraminate: alpha-2,8-sialyltransferase, GD3 synthase); sialyltransferase St8Sia I; sialytransferase St8Sia I; Sia-T; Siat8; SIAT8A; SIAT8-A; ST8 alpha-N-acetylneuraminate alpha-2,8-sialyltransferase 1; ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 1; ST8Sia I; St8sia1; ST8SiaI
Gene Symbols ST8SIA1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 49-270 of human ST8SIA1 (NP_0030251).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6489
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.