Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ST8SIA1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501734616 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-4616 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-4616 Supplier Proteintech Group Inc Supplier No. 249181AP150UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Gangliosides are membrane-bound glycosphingolipids containing sialic acid. Ganglioside GD3 is known to be important for cell adhesion and growth of cultured maligt cells. The protein encoded by this gene is a type II membrane protein that catalyzes the

Specifications

Antigen ST8SIA1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.27 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene ST8SIA1
Gene Accession No. Q64687, Q92185
Gene Alias Alpha 2,8 sialyltransferase 8A, Ganglioside GD3 synthase, Ganglioside GT3 synthase, GD3S, Sialyltransferase 8A, Sialytransferase St8Sia I, SIAT8, SIAT8 A, SIAT8A, ST8SIA1, ST8SiaI
Gene Symbols ST8SIA1
Host Species Rabbit
Immunogen ST8SIA1 Fusion Protein Ag20429
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20449, 6489
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.