Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ST8SIA3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566866
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566866 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566866 Supplier Invitrogen™ Supplier No. PA566866
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: WPFGFDPNTRE DLPYHYYDKK GTKFTTKWQE SHQLPAEFQL LYRMHGEGLT KLTLSHCA Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.

ST8SIA3 belongs to a family of sialyltransferases that form sialyl-alpha-2,8-sialyl-R linkages at the nonreducing termini of glycoconjugates.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ST8SIA3
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene St8sia3
Gene Accession No. O43173
Gene Alias AI847333; Alpha-2,8-sialyltransferase 8C; alpha-2,8-sialyltransferase III; GT3-synthase; sia-alpha-2,3-Gal-beta-1,4-GlcNAc-R:alpha 2,8-sialyltransferase; sialyltransferase 8 (alpha-2, 8-sialyltransferase) C; Sialyltransferase 8 (GT3 alpha 2,8-sialyltransferase) C; Sialyltransferase 8 (GT3 alpha 28-sialyltransferase) C; sialyltransferase 8 C; sialyltransferase 8C; sialyltransferase 8C (alpha2,3Galbeta1,4GlcNAcalpha 2,8-sialyltransferase); sialyltransferase St8Sia III; sialytransferase St8Sia III; SIAT8C; SIAT8-C; ST8 alpha-N-acetylneuraminate alpha-2,8-sialyltransferase 3; ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 3; St8sia3; ST8SiaIII
Gene Symbols St8sia3
Host Species Rabbit
Immunogen Recombinant Human ST8SIA3. Recombinant protein control fragment (Product #RP-107518).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51046
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.