Learn More
Invitrogen™ ST8SIA6 Polyclonal Antibody

Description
Sequence homology: Cow: 86%; Guinea Pig: 92%; Horse: 86%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
Specifications
Specifications
| Antigen | ST8SIA6 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 |
| Gene | ST8SIA6 |
| Gene Accession No. | P61647 |
| Gene Alias | Alpha-2,8-sialyltransferase 8F; alpha-2,8-sialyltransferase 8F variant 1; alpha-2,8-sialyltransferase 8F variant 2; alpha-2,8-sialyltransferase 8F variant 3; SIA8F; Sialyltransferase 8F; sialyltransferase 8F (alpha-2, 8-sialyltransferase); sialyltransferase St8Sia VI; sialytransferase St8Sia VI; SIAT8F; SIAT8-F; ST8 alpha-N-acetylneuraminate alpha-2,8-sialyltransferase 6; ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 6; ST8SIA6; ST8SiaVI; ST8SIA-VI |
| Gene Symbols | ST8SIA6 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.