Learn More
STA PHARMACEUTICAL US LLC STA-AM10785
Supplier:  STA PHARMACEUTICAL US LLC STAAM10785KA601GM
Encompass
WuXi TIDES 2'-OTBS G(iBu) ThioPhosphamidite - 1GR in 60 mL K&A (20-400) - S-(2-(((((2R,3R,4R,5R)-2-((bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-4-((tert-butyldimethylsilyl)oxy)-5-(2-isobutyramido-6-oxo-1,6-dihydro-9H-purin-9-yl)tetrahydrofuran-3-yl)oxy)(pyrrolidin-1-yl)phosphaneyl)thio)ethyl) benzothioate - O=C(NC(N1)=NC2=C(N=CN2[C@@H]3O[C@H](COC(C4=CC=CC=C4)(C5=CC=C(C=C5)OC)C6=CC=C(C=C6)OC)[C@@H](OP(N7CCCC7)SCCSC(C8=CC=CC=C8)=O)[C@H]3O[Si](C)(C(C)(C)C)C)C1=O)C(C)C - M.F. C54H67N6O9PS2Si, M.W. 1067.342
Catalog No. 50-286-0498