Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STAC3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501732865 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-2865 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-2865 Supplier Proteintech Group Inc Supplier No. 203921AP150UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

The Src homology 3 (SH3) domain is a highly conserved 60 amino acid protein domain that is organized into a beta-barrel fold consisting of five or six beta strands arranged as two tightly packed anti-parallel beta sheets. This domain is found in proteins that mediate assembly of specific protein complexes and interact with other proteins, specifically recognizing proline-rich regions. STAC3 (SH3 and cysteine rich domain 3) is a 364 amino acid protein containing one phorbol-ester/DAG-type zinc finger and two SH3 (Src homology 3) domains. Existing as two alternatively spliced isoforms, STAC3 maps to human chromosome 12q13.3. Human chromosome 12 encodes over 1,400 genes and comprises approximately 4.5% of the human genome. Chromosome 12 is associated with a variety of diseases and afflictions, including hypochondrogenesis, achondrogenesis, Kniest dysplasia, Noo syndrome and trisomy 12p, which causes facial developmental defects and seizure disorders.

Specifications

Antigen STAC3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.29 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene STAC3
Gene Accession No. Q8BZ71, Q96MF2
Gene Alias SH3 and cysteine rich domain 3, STAC3
Gene Symbols Stac3
Host Species Rabbit
Immunogen STAC3 Fusion Protein Ag14232
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 237611, 246329, 362895
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.