Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stachyose Tetrahydrate, MP Biomedicals™

Catalog No. 0215394690 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 mg
100 mg
500 mg
1 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-153-94690 500 mg
02-153-94601 1 mg
02-153-94680 100 mg
02-153-94691 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 02-153-94690 Supplier MP Biomedicals, Inc Supplier No. 0215394690
Add to Cart
Add to Cart

Stachyose Tetrahydrate is a prebiotic and functional oligosaccharide.

  • Promotes proliferation of lactic acid bacteria (LAB) by induction of α-galactosidase production to hydrolyze stachyose.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10094-58-3
Molecular Formula C24H42O21
Molecular Weight (g/mol) 666.58
InChI Key UQZIYBXSHAGNOE-XNSRJBNMSA-N
IUPAC Name (2S,3R,4S,5R,6R)-2-{[(2R,3R,4S,5R,6S)-6-{[(2R,3S,4S,5R,6R)-6-{[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy}-3,4,5-trihydroxyoxan-2-yl]methoxy}-3,4,5-trihydroxyoxan-2-yl]methoxy}-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO[C@H]3O[C@H](CO[C@H]4O[C@H](CO)[C@H](O)[C@H](O)[C@H]4O)[C@H](O)[C@H](O)[C@H]3O)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O

Specifications

Quantity 500 mg

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.