Learn More
Edmund Optics STAGE MICROMETER 1MM 100DIV
Supplier:  Edmund Optics NT36121
Encompass
1mm thick stage micrometer on clear glass slide with 10mm glass disc used to calibrate microscope eyepiece reticles and objective powers. 100 divisions.
Catalog No. NC0094483
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.