Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Stainless Steel Artery Forceps
SCI_ED

Catalog No. S27473 Shop All Eisco Products
Change view
Click to view available options
Tip Style:
Straight
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Tip Style
S27473 Straight
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S27473 Supplier Eisco™ Supplier No. BI0169AFS
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Feature straight tips with serrated jaws and a box joint. Stainless Steel Artery Forceps for delicate and accurate work.

  • Made from high quality stainless steel.
  • Designed for delicate and accurate work.
  • Feature straight tips with serrated jaws, box joint, and scissor handles.
  • Intended for science education purposes only.
  • Forceps should never be used on humans or other living specimen of any kind.
  • Jaw length: 1 in.
  • Non-sterile

Specifications

Product Type Artery Forceps
Autoclavable Autoclavable
Color Metallic
Material Stainless Steel
For Use With (Application) Delicate and Accurate Work
Style Serrated Jaws
Tip Style Straight
Length (English) 5 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.