Learn More
Perkin Elmer US LLC Stainless Steel ATD Prepacked Sample Tubes, Sorbent: Chromosorb 106, Pkg 10
Supplier:  Perkin Elmer US LLC N9307006
Encompass_Preferred
Sorbent:Chromasorb 106 The Chromosorb 106 is conditional at 240 °C. PerkinElmerre commends a maximum temperature of 250 °C for this sample tube with a desorption temperature of 220 °C. The tubes approximate analyte volatility range is n-C7 to n-C30 and the typical analytes for the Chromosorb 106 are a wide range of VOCs including hydrocarbons from n-C5 to n-C12. Also volatile oxygenated compounds.
Catalog No. 50-210-5283
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.