Description
- Secures vessel in precise position
- Compatible with Q3 Sample Changer
- Conductive materials to reduce electrostatic charge build-up
- Highly accurate and safe filling
Specifications
Specifications
| For Use With (Equipment) | Excellence XPR Automatic Balance |
| Size | 0 |
| Type | Stainless Steel Capsule Adapter |