Learn More
OHAUS Stainless Steel Defender 3000 Series Floor Scale
Catalog No. 01920189 Shop All OHAUS Products
Change view
Click to view available options
Model:
i-DF33XW2500C1L
i-DF33XW2500C1R
i-DF33XW5000C1L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
01-920-189 i-DF33XW5000C1L
01-920-190 i-DF33XW2500C1L
01-920-193 i-DF33XW2500C1R
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-920-189 Supplier OHAUS Supplier No. 30837606
Add to Cart
Add to Cart
Request a Quote
Defender 3000 Floor Platforms meet washdown weighing needs in industrial and commercial use. Their high-quality construction is designed to minimize flexing under heavy loads for years of dependable performance. The indicator features a bright display that can change color when in check mode.
  • 304 stainless steel, flat surface decks with 4 IP68 load cells, fixed top plate (5 mm/0.196 in. thick) with structural bracing for stiff construction.
  • Platforms available in sizes of 3 x 3 ft. and 4 x 4 ft. and capacities from 2,500 lb. to 5,000 lb. with NTEP certified 5,000e performance, allowing greater flexibility for your weighing needs.
  • Results are clearly shown on the large, bright backlit LCD display.
  • In checkweighing mode, display changes color (yellow/green/red) to alert users when within the target weight range.
TRUSTED_SUSTAINABILITY
Specifications
Capacity 2500 kg
Type Scale
Display Type LCD
Readability 0.5 kg
Legal for Trade Class III
Model i-DF33XW5000C1L