Learn More
Description
Includes
Two stainless-steel shelves, desiccant tray
Key Features
- Polished stainless steel with 0.187 in. thick (4.8mm) glass
- Airtight door with neoprene gasket, full-height hinges and double-action bar locks
- Fixed wire shelf brackets every 0.5 in. (13mm)
Standard Size
- Glass on door and sides
- Tray holds 16 oz. (454g) of desiccant
- Weighs 23 lb. (10kg)
Large Size
- Glass on door and back, stainless-steel sides
- Shelf dimensions: 13.375 x 18 in. (34 x 46cm)
- Weighs 57 lb. (26kg)
Warranty and Services
Two years
Specifications
Specifications
| Material | Stainless Steel |
| Product Type | Desiccator Cabinet |
| For Use With (Application) | Storing reagents or soil samples, drying biological specimens or storage sludge samples |
| Includes | Airtight door with neoprene gasket, full-height hinges, double-action bar locks, two stainless steel shelves and desiccant tray |
| Size | Standard |
| Dimensions (L x W x H) | 12.25 x 12.5 x 12.25 in. (31.1 x 31.7 x 31 cm) |
| Length (English) | 12.25 in. |
| Length (Metric) | 31.1 cm |
| Width (English) | 12.5 in. |
| Width (Metric) | 31.7 cm |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

