Learn More
United Scientific™ Stainless Steel Double Adjustable Burette Clamp
SDPSCI_ED
Catalog No. S00328 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Color:
Silver
Catalog No. Color
S00328 Silver
1 options
This item is not returnable. View return policy
Catalog No. S00328 Supplier United Scientific™ Supplier No. UNDBBR01
Add to Cart
Add to Cart
This item is not returnable. View return policy
Versatile clamp that features two separate sets of adjustable jaws for securing two burettes or any type of cylindrical tubes simultaneously.
  • The PVC-coated jaws can swivel 90 degrees so they can be next to each other or separated
  • The jaws are rounded to provide a better grip on round tubes, and can be swiveled and locked at any angle
  • Plated brass nuts enable easy positioning of the clamps
  • The all-stainless steel jaws open from 15 to 45 mm (0.5 to 1.75 in.)
  • The die-cast aluminum bosshead attaches to rods up to 13 mm (0.5 in.) O.D
Specifications
Type Double Clamp
Material Alloy, Stainless Steel
Color Silver
Jaw Type 2-Prong Single Adjust