Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stainless Steel flake, -150 mesh, 1.2 micron thick, Type 316-L

Catalog No. AA42486A1
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA42486A1 1 kg
AA4248630 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA42486A1 Supplier Thermo Scientific Chemicals Supplier No. 042486A1
Add to Cart

Stainless Steel is most commonly utilized for wire cloth weaving. Typical wire cloth applications include use with chemicals, food products, pharmaceuticals and exposure to moisture.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Stainless Steel flake, Type 316-L
Name Note 1.2 micron thick
Avg. Mol. Wt. or Mol. Wt. Range Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%
Quantity 1 kg
Mesh Size −150 Mesh
Form Flake
MDL Number MFCD00148499
Solubility Information Insoluble in water.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.