Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stainless Steel foil, 0.5mm (0.02in) thick, Type 304

Catalog No. AA41585FF
Change view
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
100 x 100 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA41585FF 25 x 25 mm
AA41585FI 50 x 50 mm
AA41585GH 100 x 100 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA41585FF Supplier Thermo Scientific Chemicals Supplier No. 041585FF
Add to Cart

Fe:Cr:Ni; 70:19:11 wt%

It is most commonly utilized for wire cloth weaving. Typical wire cloth applications include use with chemicals, food products, pharmaceuticals and exposure to moisture.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Color Silver
Chemical Name or Material Stainless Steel
Appearance Silver -colored foil
Quantity 25 x 25 mm
Odor Odorless
Form Foil
Molecular Formula Fe:Cr:Ni; 70:19:11 wt%
MDL Number MFCD00148499
Solubility Information It is insoluble in water.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.