Learn More
MilliporeSigma™ Supelco™ Stainless Steel Graphite Ferrules
Catalog No. 111004640 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Packaging Quantity:
4/Pk.
Catalog No. Packaging Quantity
11-100-4640 4/Pk.
1 options
Catalog No. 11-100-4640 Supplier MilliporeSigma™ Supelco™ Supplier No. 23311
Add to Cart
Add to Cart
Graphite ferrules and stainless steel spacers for use with 5 mm columns and liners in Shimadzu GCs.
TRUSTED_SUSTAINABILITY
Specifications
Packaging Quantity 4/Pk.
Material Stainless Steel