Learn More
MilliporeSigma™ Supelco™ Stainless Steel HPLC Fittings Ferrule for for 1/16 in. Tubing
Catalog No. 111016284 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Packaging Quantity:
10/Pk.
Catalog No. Packaging Quantity
11-101-6284 10/Pk.
1 options
Catalog No. 11-101-6284 Supplier MilliporeSigma™ Supelco™ Supplier No. 58762
The stainless steel ferrule is used for connecting most HPLC columns to a 1/16 in. tubing system.
TRUSTED_SUSTAINABILITY
Specifications
Packaging Quantity 10/Pk.
Material Stainless Steel