Learn More
JDM SCIENTIFIC LLC Stainless steel round for insertion into pipette tip boxes
Supplier:  JDM SCIENTIFIC LLC 01167
Encompass_Preferred
The round made of magnetic stainless steel can be inserted into a pipette tip box, thereby enabling the pipette tip box to be held securely on the holder for pipette tip boxes.
Catalog No. 50-255-8542
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.