Learn More
Diversified Spaces™ Stainless-Steel Sink
SCI_ED
Catalog No. S37872 Shop All Diversified Spaces Products
Change view
Click to view available options
Dimensions (D x W x H):
30 x 60 x 35.75 in.
30 x 72 x 35.75 in.
Shelf Type:
Galvanized undershelf
Style:
Left side sink
Right side sink
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (D x W x H) Shelf Type Style
S37872 30 x 60 x 35.75 in. Galvanized undershelf Left side sink
S37873 30 x 60 x 35.75 in. Galvanized undershelf Right side sink
S37875 30 x 72 x 35.75 in. Galvanized undershelf Right side sink
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S37872 Supplier Diversified Spaces™ Supplier No. 250495
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
With Stainless steel bowl with holes for a deck-mounted faucet
  • Includes drain basket
  • 1 x 2 in. tubular channels run left to right under work surface
  • 1 x 2 in. hat channel connects tubular channels
  • Rounded edges
  • Shelves can be adjusted for preferred height
Specifications
Product Type Stainless Steel Sink
Style Left side sink
Includes drain basket
Dimensions (D x W x H) 30 x 60 x 35.75 in.
Shelf Type Galvanized undershelf