Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stainless Steel spheres, 3mm (0.12 in.) dia., Type 302, Thermo Scientific™

Catalog No. AA42017KM
Change view
Click to view available options
25 pieces
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AA42017KM 25 pieces
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA42017KM Supplier Thermo Scientific Chemicals Supplier No. 42017KM

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Stainless Steel spheres, Type 302
Name Note 3mm (0.12 in.) dia.
Avg. Mol. Wt. or Mol. Wt. Range Fe:Cr:Ni:Mn:Si; 70:18:9:2:1 wt%
MDL Number MFCD00148499
Quantity 25 pc.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.