Learn More
United Scientific™ Stainless Steel Table Clamp
SDPSCI_ED
Catalog No. S03072 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Includes:
No Rod
Rod
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
S03072 No Rod
S03071 Rod
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Prop 65
Catalog No. S03072 Supplier United Scientific™ Supplier No. SSTBLCLMP
Add to Cart
Add to Cart
Prop 65
Stainless Steel Table Clamp saves space when there isn't room for a large support base.
Stainless steel support clamp for lab tables. Hold support rods securely to any table edge up to 2.2 inches thick. The set screw allows for easy adjustments to hold support rods at various heights.
  • Holds rods up to 0.5 inches in diameter
  • Fits table edges up to 2.2 inches.
  • Solid stainless steel, 0.6 pounds.
  • Available with and without a support rod.
Specifications
Type Adjustable Clamp
Material Nickel-plated
Includes No Rod
Color Silver
Jaw Type Single Adjustment