Learn More
Perkin Elmer US LLC Stainless Steel TD tubes Conditioned
Supplier:  Perkin Elmer US LLC N9306277
Encompass_Preferred
Sorbent: Soil Vapor Intrusion Our new multi-bed construction extends the hydrocarbon range past naphthalene while retaining the lighter components, enabling larger sample volumes, hence, enhancing detection limits. Unique design that meets the challenges and criteria of the EPA regulations for air monitoring. Enhanced recoveries, even of highly contaminated sites From chloromethane through diesel range hydrocarbons After the analysis, tubes are clean and ready for re-sampling reducing costs.
Catalog No. 50-210-4672
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.