Learn More
Bio-Serv™ Stainless Steel Twist Clip
SDP
Catalog No. 14727038
Encompass
Change view
Click to view available options
Autoclavable:
Autoclavable
Catalog No. Autoclavable
14-727-038 Autoclavable
1 options
Catalog No. 14-727-038 Supplier Bio-Serv™ Supplier No. 9022
  • Made of stainless steel
  • Use to suspend various enrichment devices
  • Autoclavable
  • Cagewasher safe
  • 2.25 in. long x .375 in. inside width x .125 in. thickness
Specifications
Autoclavable Autoclavable
Material Stainless Steel
For Use With (Application) To suspend various enrichment devices
Thickness (English) Wall 0.125 in.
Length (English) 2.25 in.
Width (English) 0.375 in.