Learn More
Vikan™ Stainless Steel Undercarriage
Catalog No. 17227658 Shop All Remco Products
Change view
Click to view available options
Color:
Steel
Catalog No. Color
17-227-658 Steel
1 options
Catalog No. 17-227-658 Supplier Vikan™ Supplier No. 6906
Add to Cart
Add to Cart
Stainless Steel Undercarriage is commonly used for meat, seafood, poultry, commercial fishing, confectionery, bakery, and grain applications.
  • Tall
  • Stainless steel undercarriage
  • Fatures caster wheels that are impervious to meat fat and bearings equipped with grease fittings
  • The undercarriage fits tubs 6905, 6901, 6911, and 6915 to provide an easy method of moving large amounts of product through a production facility
  • This model includes a built-in stop that allows the tub to be tipped and emptied
Specifications
Product Type Undercarriage
Color Steel
Material Stainless Steel
Quantity 1
Dimensions (L x W x H) 41.1 x 27.3 x 19.1 in.