Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Stains-All, Cationic Carbocyanine Dye, >95% Purity, 1 g

Catalog No. 502704764
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-4764 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-4764 Supplier Abcam Supplier No. AB1462781G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cationic carbocyanine dye used for differential staining of nucleic acids and proteins in polyacrylamide gels.

  • Molecular weight: 559.58 g/mol
  • Purity: >95%
  • Description: Cationic carbocyanine dye used for differential staining of nucleic acids and proteins. RNA stains blue/purple, DNA stains blue, and proteins stain red.
  • Applications: Visualization and identification of proteins on polyacrylamide gels; differentiates between highly acidic proteins (blue) and less acidic proteins (pink).
  • Performance: Staining of Ca2+-binding proteins in muscle extracts is 50 to 100% more intense than Coomassie blue staining. Staining intensity on polyacrylamide gels can be increased with the addition of silver nitrate.
  • Storage: Store at ambient temperature.

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Shipping Condition Ambient
Substrate Properties Chemical Substrate
Target Molecule RNA, DNA, Proteins, Ca2+-binding proteins
Form Powder
Content And Storage Store at ambient temperature.
Buffer 100% Stains-All
Detection Location In-Gel Detection
Detection Method Colorimetric
Formulation 100% Stains-All
Sample Type Muscle extracts
Quantity 1 g
For Use With (Application) Differential staining of nucleic acids and proteins, Protein visualization and identification on polyacrylamide gels
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.